Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044492.1 | NP_001037957.1 | 3 - 4 | 376 | 95 | |
XM_012963095.2 | XP_012818549.1 | 3 - 4 | 488 | 95 | ||
XM_018093927.1 | XP_017949416.1 | 2 - 3 | 369 | 95 | ||
GenBank mRNA | BC169138.1 | - | 3 - 4 | 371 | 95 | |
CR760495.2 | - | 3 - 4 | 376 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |