Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044493.1 | NP_001037958.1 | 7 - 8 | 922 | 71 | |
XM_012966681.2 | XP_012822135.1 | 8 - 9 | 1350 | 71 | ||
GenBank mRNA | BC121360.1 | - | 8 - 9 | 1321 | 71 | |
BC170738.1 | - | 7 - 8 | 908 | 71 | ||
BC170742.1 | - | 7 - 8 | 908 | 71 | ||
CR760664.2 | - | 7 - 8 | 922 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |