Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044503.1 | NP_001037968.1 | 3 - 4 | 363 | 100 | |
XM_012957128.1 | XP_012812582.1 | 4 - 5 | 697 | 100 | ||
XM_012957129.2 | XP_012812583.1 | 4 - 5 | 407 | 100 | ||
XM_012957130.1 | XP_012812584.1 | 4 - 5 | 383 | 100 | ||
GenBank mRNA | CR761596.2 | - | 3 - 4 | 363 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Xenopus tropicalis Probe spans exons |