Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: szl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044506.1 | NP_001037971.1 | 1 - 2 | 280 | 85 | |
GenBank mRNA | CR761702.2 | - | 1 - 2 | 280 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Xenopus tropicalis Probe spans exons |