Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044511.1 | NP_001037976.1 | 3 - 4 | 374 | 82 | |
XM_012961250.2 | XP_012816704.1 | 3 - 4 | 297 | 82 | ||
GenBank mRNA | BC135182.1 | - | 3 - 4 | 360 | 82 | |
CR761848.2 | - | 3 - 4 | 374 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |