Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044516.1 | NP_001037981.1 | 5 - 6 | 814 | 72 | |
XM_012964182.2 | XP_012819636.1 | 5 - 6 | 906 | 72 | ||
XM_018093983.1 | XP_017949472.1 | 5 - 6 | 801 | 72 | ||
GenBank mRNA | CR761942.2 | - | 5 - 6 | 814 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |