Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044517.1 | NP_001037982.1 | 4 - 5 | 1150 | 96 | |
GenBank mRNA | CR761976.1 | - | 4 - 5 | 1150 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |