Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045580.1 | NP_001039045.1 | 766 | 75 | ||
GenBank mRNA | CR848392.2 | - | 766 | 75 |