Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c8orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045584.1 | NP_001039049.1 | 2 - 3 | 271 | 77 | |
GenBank mRNA | BC161270.1 | - | 2 - 3 | 198 | 77 | |
CR848448.2 | - | 2 - 3 | 271 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |