Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045608.1 | NP_001039073.1 | 3 - 4 | 334 | 72 | |
XM_012953172.2 | XP_012808626.1 | 3 - 4 | 417 | 72 | ||
XM_012953173.1 | XP_012808627.1 | 3 - 4 | 397 | 72 | ||
XM_012953174.2 | XP_012808628.1 | 3 - 4 | 387 | 72 | ||
GenBank mRNA | BC135708.1 | - | 2 - 3 | 211 | 72 | |
CR855609.1 | - | 3 - 4 | 334 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |