Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045612.2 | NP_001039077.1 | 4 - 5 | 939 | 65 | |
XM_018096288.1 | XP_017951777.1 | 3 - 4 | 1058 | 65 | ||
GenBank mRNA | BC123083.1 | - | 4 - 5 | 939 | 65 | |
CR855644.1 | - | 4 - 5 | 766 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |