Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045612.2 | NP_001039077.1 | 7 - 8 | 1212 | 70 | |
XM_012968543.2 | XP_012823997.1 | 6 - 7 | 1235 | 70 | ||
XM_018096288.1 | XP_017951777.1 | 6 - 7 | 1331 | 70 | ||
GenBank mRNA | BC123083.1 | - | 7 - 8 | 1212 | 70 | |
CR855644.1 | - | 7 - 8 | 1039 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |