Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045623.1 | NP_001039088.1 | 5 - 6 | 655 | 84 | |
GenBank mRNA | BC135635.1 | - | 5 - 6 | 663 | 84 | |
BC160791.1 | - | 5 - 6 | 661 | 84 | ||
CR926372.2 | - | 5 - 6 | 655 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |