Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045627.1 | NP_001039092.1 | 6 - 7 | 613 | 81 | |
GenBank mRNA | BC154054.1 | - | 6 - 7 | 579 | 81 | |
CR942347.2 | - | 6 - 7 | 613 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |