Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tor1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045631.1 | NP_001039096.1 | 2 - 3 | 504 | 63 | |
GenBank mRNA | BC160421.1 | - | 2 - 3 | 508 | 63 | |
CR942373.2 | - | 2 - 3 | 504 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |