Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aes | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045637.1 | NP_001039102.1 | 5 - 6 | 583 | 71 | |
XM_012964320.2 | XP_012819774.1 | 5 - 6 | 664 | 71 | ||
XM_018094049.1 | XP_017949538.1 | 5 - 6 | 672 | 71 | ||
XM_018094052.1 | XP_017949541.1 | 5 - 6 | 671 | 71 | ||
XM_018094056.1 | XP_017949545.1 | 4 - 5 | 578 | 71 | ||
GenBank mRNA | BC118730.1 | - | 5 - 6 | 601 | 71 | |
BC135945.1 | - | 5 - 6 | 566 | 71 | ||
CR942389.2 | - | 5 - 6 | 583 | 71 | ||
GU014561.1 | - | 5 - 6 | 606 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |