Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045650.1 | NP_001039115.1 | 4 - 5 | 561 | 102 | |
XM_018096290.1 | XP_017951779.1 | 4 - 5 | 621 | 102 | ||
XM_018096291.1 | XP_017951780.1 | 4 - 5 | 2061 | 102 | ||
GenBank mRNA | BC160407.1 | - | 4 - 5 | 599 | 102 | |
CR942445.2 | - | 4 - 5 | 561 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |