Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: admp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045692.1 | NP_001039157.1 | 1 - 2 | 330 | 84 | |
GenBank mRNA | CR942593.2 | - | 1 - 2 | 330 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |