Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045706.1 | NP_001039171.1 | 4 - 5 | 617 | 65 | |
XM_012964429.2 | XP_012819883.1 | 3 - 4 | 907 | 65 | ||
XM_012964436.2 | XP_012819890.1 | 3 - 4 | 918 | 65 | ||
XM_012964442.2 | XP_012819896.1 | 3 - 4 | 915 | 65 | ||
XM_012964450.2 | XP_012819904.1 | 4 - 5 | 672 | 65 | ||
XM_012964456.2 | XP_012819910.1 | 4 - 5 | 642 | 65 | ||
XM_018094098.1 | XP_017949587.1 | 3 - 4 | 920 | 65 | ||
GenBank mRNA | CR761735.2 | - | 4 - 5 | 617 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |