Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045712.1 | NP_001039177.1 | 2 - 3 | 320 | 93 | |
XM_012957176.2 | XP_012812630.1 | 4 - 5 | 644 | 93 | ||
XM_012957177.2 | XP_012812631.1 | 3 - 4 | 499 | 93 | ||
XM_018091322.1 | XP_017946811.1 | 4 - 5 | 640 | 93 | ||
XM_018091323.1 | XP_017946812.1 | 3 - 4 | 343 | 93 | ||
GenBank mRNA | CR762036.2 | - | 2 - 3 | 320 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |