Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stoml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045728.1 | NP_001039193.1 | 2 - 3 | 409 | 71 | |
XM_018092141.1 | XP_017947630.1 | 2 - 3 | 581 | 71 | ||
GenBank mRNA | CR926277.2 | - | 2 - 3 | 409 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |