Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045736.1 | NP_001039201.1 | 4 - 5 | 1324 | 64 | |
GenBank mRNA | CR942394.2 | - | 4 - 5 | 1324 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |