Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045745.1 | NP_001039210.1 | 13 - 14 | 2579 | 86 | |
XM_012968564.2 | XP_012824018.1 | 14 - 15 | 2688 | 86 | ||
XM_012968566.2 | XP_012824020.1 | 11 - 12 | 2461 | 86 | ||
XM_018096295.1 | XP_017951784.1 | 13 - 14 | 2538 | 86 | ||
GenBank mRNA | CR942488.2 | - | 13 - 14 | 2579 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |