Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045750.1 | NP_001039215.1 | 3 - 4 | 227 | 81 | |
XM_018093934.1 | XP_017949423.1 | 3 - 4 | 1042 | 81 | ||
GenBank mRNA | BC135657.1 | - | 3 - 4 | 215 | 81 | |
CR942549.2 | - | 3 - 4 | 227 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |