Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045774.1 | NP_001039239.1 | 4 - 5 | 453 | 83 | |
XM_012970515.2 | XP_012825969.1 | 4 - 5 | 521 | 83 | ||
GenBank mRNA | BC158417.1 | - | 4 - 5 | 483 | 83 | |
CR942753.2 | - | 4 - 5 | 453 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |