Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045784.1 | NP_001039249.1 | 1306 | 54 | ||
GenBank mRNA | BC168785.1 | - | 10 - 11 | 996 | 54 | |
CR942792.2 | - | 1306 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |