Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045803.1 | NP_001039268.1 | 3 - 4 | 520 | 63 | |
XM_012964858.2 | XP_012820312.1 | 4 - 5 | 731 | 63 | ||
XM_012964859.2 | XP_012820313.1 | 5 - 6 | 869 | 63 | ||
XM_018094689.1 | XP_017950178.1 | 5 - 6 | 797 | 63 | ||
GenBank mRNA | BC135988.1 | - | 5 - 6 | 847 | 63 | |
CR942790.2 | - | 3 - 4 | 520 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |