Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078710.1 | NP_001072178.1 | 9 - 10 | 4914 | 107 | |
XM_012957215.2 | XP_012812669.1 | 9 - 10 | 4690 | 107 | ||
XM_018091331.1 | XP_017946820.1 | 8 - 9 | 5600 | 107 | ||
XM_018091332.1 | XP_017946821.1 | 8 - 9 | 4585 | 107 | ||
GenBank mRNA | DQ886532.1 | - | 9 - 10 | 4914 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons |