Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078733.1 | NP_001072201.1 | 5 - 6 | 478 | 88 | |
XM_012957216.2 | XP_012812670.1 | 5 - 6 | 550 | 88 | ||
GenBank mRNA | BC118751.1 | - | 5 - 6 | 478 | 88 | |
BC158969.1 | - | 5 - 6 | 470 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |