Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078743.1 | NP_001072211.1 | 2 - 3 | 272 | 82 | |
XM_012961289.2 | XP_012816743.1 | 2 - 3 | 294 | 82 | ||
GenBank mRNA | BC118779.1 | - | 2 - 3 | 272 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |