Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078745.1 | NP_001072213.1 | 9 - 10 | 1095 | 101 | |
XM_012964876.2 | XP_012820330.1 | 10 - 11 | 3245 | 101 | ||
XM_012964879.2 | XP_012820333.1 | 8 - 9 | 924 | 101 | ||
GenBank mRNA | BC118782.1 | - | 9 - 10 | 1095 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |