Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078751.1 | NP_001072219.1 | 1 - 2 | 500 | 95 | |
XM_012961290.2 | XP_012816744.1 | 1 - 2 | 1073 | 95 | ||
GenBank mRNA | BC118797.1 | - | 1 - 2 | 500 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |