Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078786.1 | NP_001072254.1 | 10 - 11 | 1347 | 73 | |
XM_018094323.1 | XP_017949812.1 | 7 - 8 | 699 | 73 | ||
GenBank mRNA | BC121219.1 | - | 10 - 11 | 1347 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |