Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078793.1 | NP_001072261.1 | 3 - 4 | 1342 | 66 | |
XM_018092157.1 | XP_017947646.1 | 3 - 4 | 1068 | 66 | ||
GenBank mRNA | BC121249.1 | - | 3 - 4 | 1342 | 66 | |
CT027965.3 | - | 3 - 4 | 1340 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |