Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078804.1 | NP_001072272.1 | 688 | 65 | ||
GenBank mRNA | BC121276.1 | - | 688 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |