Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 13 - 14 | 2776 | 69 | |
XM_012952548.2 | XP_012808002.1 | 13 - 14 | 3330 | 69 | ||
GenBank mRNA | BC121308.1 | - | 13 - 14 | 2776 | 69 | |
CR926259.1 | - | 1 - 2 | 143 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |