Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078831.1 | NP_001072299.1 | 1078 | 74 | ||
GenBank mRNA | BC121344.1 | - | 1078 | 74 | ||
CR855465.2 | - | 1082 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |