Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phyhipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078853.1 | NP_001072321.1 | 2 - 3 | 731 | 105 | |
XM_012966732.1 | XP_012822186.1 | 2 - 3 | 319 | 105 | ||
XM_018095430.1 | XP_017950919.1 | 2 - 3 | 305 | 105 | ||
GenBank mRNA | BC121389.1 | - | 2 - 3 | 731 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |