Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078931.2 | NP_001072399.2 | 3 - 4 | 941 | 92 | |
XM_012959240.2 | XP_012814694.1 | 3 - 4 | 1422 | 92 | ||
GenBank mRNA | BC121602.1 | - | 3 - 4 | 92 | ||
BC160471.1 | - | 3 - 4 | 979 | 92 | ||
CU075314.1 | - | 3 - 4 | 932 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |