Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078947.1 | NP_001072415.1 | 2 - 3 | 307 | 96 | |
XM_012965232.2 | XP_012820686.1 | 2 - 3 | 841 | 96 | ||
GenBank mRNA | BC121640.1 | - | 2 - 3 | 307 | 96 | |
CU024960.1 | - | 2 - 3 | 318 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |