Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078964.1 | NP_001072432.1 | 2 - 3 | 239 | 98 | |
XM_012957263.2 | XP_012812717.1 | 3 - 4 | 330 | 98 | ||
XM_012957264.2 | XP_012812718.1 | 3 - 4 | 458 | 98 | ||
XM_012957267.2 | XP_012812721.1 | 3 - 4 | 270 | 98 | ||
XM_018091341.1 | XP_017946830.1 | 4 - 5 | 378 | 98 | ||
GenBank mRNA | BC121678.1 | - | 2 - 3 | 239 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |