Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079014.1 | NP_001072482.1 | 9 - 10 | 1435 | 84 | |
GenBank mRNA | BC121907.1 | - | 9 - 10 | 1435 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |