Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079206.1 | NP_001072674.1 | 2 - 3 | 377 | 90 | |
GenBank mRNA | BC124001.1 | - | 2 - 3 | 377 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |