Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079206.1 | NP_001072674.1 | 4 - 5 | 672 | 85 | |
XM_012957298.2 | XP_012812752.1 | 3 - 4 | 899 | 85 | ||
GenBank mRNA | BC124001.1 | - | 4 - 5 | 672 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |