Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079219.1 | NP_001072687.1 | 3 - 4 | 486 | 74 | |
XM_012965802.1 | XP_012821256.1 | 3 - 4 | 541 | 74 | ||
XM_012965806.2 | XP_012821260.1 | 3 - 4 | 544 | 74 | ||
XM_018094605.1 | XP_017950094.1 | 3 - 4 | 235 | 74 | ||
GenBank mRNA | BC124020.1 | - | 3 - 4 | 486 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |