Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079231.1 | NP_001072699.1 | 5 - 6 | 1040 | 82 | |
XM_012965809.2 | XP_012821263.1 | 5 - 6 | 1097 | 82 | ||
GenBank mRNA | BC124042.1 | - | 5 - 6 | 1040 | 82 | |
BC160577.1 | - | 5 - 6 | 986 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |