Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079236.2 | NP_001072704.1 | 8 - 9 | 1447 | 84 | |
XM_012965810.2 | XP_012821264.1 | 9 - 10 | 1584 | 84 | ||
XM_012965811.2 | XP_012821265.1 | 10 - 11 | 1687 | 84 | ||
XM_012965812.2 | XP_012821266.1 | 9 - 10 | 1534 | 84 | ||
XM_012965813.2 | XP_012821267.1 | 8 - 9 | 1506 | 84 | ||
GenBank mRNA | BC124048.1 | - | 8 - 9 | 1448 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |