Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079273.2 | NP_001072741.1 | 2 - 3 | 370 | 74 | |
NM_001358944.1 | NP_001345873.1 | 2 - 3 | 370 | 74 | ||
NM_001358946.1 | NP_001345875.1 | 2 - 3 | 370 | 74 | ||
GenBank mRNA | BC125729.1 | - | 2 - 3 | 370 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |