Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079294.1 | NP_001072762.1 | 4 - 5 | 496 | 91 | |
GenBank mRNA | BC125794.1 | - | 4 - 5 | 496 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |