Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079303.1 | NP_001072771.1 | 4 - 5 | 918 | 89 | |
XM_012957324.2 | XP_012812778.1 | 4 - 5 | 1129 | 89 | ||
GenBank mRNA | BC125809.1 | - | 4 - 5 | 918 | 89 | |
DQ658156.1 | - | 2 - 3 | 690 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |